accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17476-04-9

LiAlH3CCH3CH3OCH3CH3CH3OCH3CH3CH3O

CAS RN 17476-04-9

IUPAC Name: aluminium(3+) tris(2-methylpropan-2-olate) lithium
Synonyms: aluminium(3+) tris(2-methylpropan-2-olate) lithium
Molecular Weight (g/mol): 253.27
Molecular Formula: C12H27AlLiO3
InChi Key: HTBVGZAVHBZXMS-UHFFFAOYSA-N
SMILES: [Li].[Al+3].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)