CAS RN 166881-42-1
CAS RN 166881-42-1
IUPAC Name:
2-{[(2,4-dimethoxyphenyl)methyl]({[(9H-fluoren-9-yl)methoxy]carbonyl})amino}acetic acid
Synonyms:
{[(2,4-dimethoxyphenyl)methyl][(9H-fluoren-9-ylmethoxy)carbonyl]amino}acetic acid
Molecular Weight (g/mol):
447.49
Molecular Formula:
C26H25NO6
InChi Key:
UIDQSTVPYKMCEY-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=C(CN(CC(O)=O)C(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C=C1
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc- Dmb Gly-OH, Advanced ChemTech
CAS: 166881-42-1 Molecular Formula: C26H25NO6 Molecular Weight (g/mol): 447.49 MDL Number: MFCD08064316 InChI Key: UIDQSTVPYKMCEY-UHFFFAOYSA-N IUPAC Name: 2-{[(2,4-dimethoxyphenyl)methyl]({[(9H-fluoren-9-yl)methoxy]carbonyl})amino}acetic acid SMILES: COC1=CC(OC)=C(CN(CC(O)=O)C(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C=C1