CAS RN 124495-18-7
CAS RN 124495-18-7
IUPAC Name:
5,7-dichloro-4-(4-fluorophenoxy)quinoline
Synonyms:
5,7-dichloro-4-(4-fluorophenoxy)quinoline
Molecular Weight (g/mol):
308.13
Molecular Formula:
C15H8Cl2FNO
InChi Key:
WRPIRSINYZBGPK-UHFFFAOYSA-N
SMILES:
FC1=CC=C(OC2=C3C(Cl)=CC(Cl)=CC3=NC=C2)C=C1
1
–
1
of
1
results
1
Quinoxyfen 98.0+%, TCI America™
CAS: 124495-18-7 Molecular Formula: C15H8Cl2FNO Molecular Weight (g/mol): 308.13 MDL Number: MFCD03265638 InChI Key: WRPIRSINYZBGPK-UHFFFAOYSA-N Synonym: quinoxyfen,5,7-dichloro-4-4-fluorophenoxy quinoline,legend,quinoline, 5,7-dichloro-4-4-fluorophenoxy,unii-ppc78j1vcw,quinoxyfen iso:bsi:ansi,ppc78j1vcw,5,7-dichloro-4-quinolyl 4-fluorophenyl ether,acmc-20mr2i,dsstox_cid_14881 PubChem CID: 3391107 ChEBI: CHEBI:82040 IUPAC Name: 5,7-dichloro-4-(4-fluorophenoxy)quinoline SMILES: FC1=CC=C(OC2=C3C(Cl)=CC(Cl)=CC3=NC=C2)C=C1