accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1121-76-2

ONCl

CAS RN 1121-76-2

IUPAC Name: 4-chloropyridin-1-ium-1-olate
Synonyms: 4-chloropyridine N-oxide
Molecular Weight (g/mol): 129.54
Molecular Formula: C5H4ClNO
InChi Key: DPJVRASYWYOFSJ-UHFFFAOYSA-N
SMILES: [O-][N+]1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)