accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106-69-4

HOOHOH

CAS RN 106-69-4

IUPAC Name: hexane-1,2,6-triol
Synonyms: 1,2,6-hexanetriol
Molecular Weight (g/mol): 134.18
Molecular Formula: C6H14O3
InChi Key: ZWVMLYRJXORSEP-UHFFFAOYNA-N
SMILES: OCCCCC(O)CO
Molecular Weight (g/mol) 
  • (2)
  • (16)
Percent Purity 
  • (2)
  • (2)
  • (12)
  • (2)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (4)
  • (2)
  • (2)
Grade 
  • (2)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (8)
  • (4)
  • (4)
  • (12)
  • (2)
  • (16)
  • (2)
  • (2)
  • (12)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (8)
  • (2)
  • (2)
  • (2)