Assay Details
| Gene Symbol: mRpS35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079169.3 | NP_523893.1 | 1 - 2 | 230 | 78 | |
| GenBank mRNA | BT016117.1 | - | 1 - 2 | 208 | 78 | |
More Information
Set Membership: |
|

mRpS35
Genomic Map