Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZWINT Antibody, Novus Biologicals™
SDP

Catalog No. NB1002364 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002364 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002364 Supplier Novus Biologicals Supplier No. NB1002364
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZWINT Polyclonal specifically detects ZWINT in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen ZWINT
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation 2-5 μg/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O95229
Gene Alias C20orf164, chromosome 20 open reading frame 164, dJ337O18.7, zinc finger SWIM domain-containing protein 3, zinc finger, SWIM domain containing 3, zinc finger, SWIM-type containing 3
Gene Symbols ZWINT
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 225 and the C-terminus (residue 277) of human ZW10 Interactor-1 using the numbering given in entry NP_008988.2 (GeneID 11130).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11130
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.