Learn More
Zymo Research Corporation Quick-RNA Viral 96™ Kit (2 x 96 Preps)
Supplier: Zymo Research Corporation R1040
The Quick-RNA Viral 96 Kit provides for rapid, large-scale isolation of viral RNA from plasma, serum, cell culture media, cellular suspensions, urine, blood, saliva and any other biological samples stored in DNA/RNA Shield. DNA/RNA Shield ensures nucleic acid stability during sample storage/transport at ambient temperatures. The reagent effectively lyses cells and inactivates nucleases and infectious agents (virus). The kit features a specialized buffer system that facilitates complete viral particle lysis for efficient RNA isolation from samples containing enteroviruses, rhinoviruses, coronaviruses, HIV, HCV, influenza A virus, flaviviruses, measles virus, parainfluenza virus, parvovirus (a ssDNA virus), etc. Viral RNA is bound to the matrix in each well of the spin-plate, washed and eluted. The isolated high-quality viral RNA is ready for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/PCR detection.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.