Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZOLL™ Medical Batteries for AED Plus™ Defibrillators
Shop All Zoll Medical Corporation Products
Click to view available options
Product Type:
Battery, Type 123 Lithium
Description
- For use with ZOLL AED Plus semi-automatic and fully-automatic
- Shelf life of 5 years
Specifications
Specifications
| Product Type | Battery, Type 123 Lithium |
| Material | Lithium Type 123 |
| For Use With (Equipment) | AED Plus Defibrillator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction