Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc neodecanoate, Zn 17.9-18.2%

Catalog No. AA4131230
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4131230 250 g
AA41312A1 1 kg
AA41312A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4131230 Supplier Thermo Scientific Chemicals Supplier No. 04131230
Only null left
Add to Cart

CAS: 27253-29-8 | C20H40O4Zn | 409.916 g/mol

Zinc neodecanoate is used as a catalyst, lubricant additive and drier. Further it is used in the production of rubber and plastic products. In addition it serves as a polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27253-29-8
Molecular Formula C20H40O4Zn
Molecular Weight (g/mol) 409.916
MDL Number MFCD00014384
InChI Key VADFQXONMAETJP-UHFFFAOYSA-N
Synonym zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
PubChem CID 117064711
IUPAC Name 7,7-dimethyloctanoic acid;zinc
SMILES CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]

Specifications

Color Brown
Flash Point 102°C (216°F)
Quantity 250 g
Solubility Information Immiscible with water.
Formula Weight 407.89
Physical Form Liquid
Chemical Name or Material Zinc neodecanoate
EINECSNumber 248-370-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.