Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113144
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5113144 100 μL
1 options

Catalog No. PIPA5113144

Supplier: Invitrogen™ PA5113144

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZDHHC12 Polyclonal Antibody for IHC (P), ELISA

ZDHHC12 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein palmitoylation; protein targeting to membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC12
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.184 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZDHHC12
Gene Accession No. Q96GR4
Gene Alias 1190004A01Rik; DHHC-12; membrane-associated DHHC12 zinc finger protein; probable palmitoyltransferase ZDHHC12; PSEC0008; ZDHHC12; Zinc finger DHHC domain-containing protein 12; zinc finger DHHC-type containing 12; zinc finger DHHC-type palmitoyltransferase 12; zinc finger protein 400; zinc finger, DHHC domain containing 12; zinc finger, DHHC-type containing 12; ZNF400
Gene Symbols ZDHHC12
Host Species Rabbit
Immunogen Recombinant Human Probable palmitoyltransferase ZDHHC12 protein (121-197AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84885
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.