Learn More
Teknova YPD Broth. 1000mL, Sterile.
Supplier: Teknova Y5000
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.