Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ymer Antibody, Novus Biologicals™
SDP

Catalog No. NB1001542 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
Catalog No. Quantity
NB1001542 0.1 mg
1 options

Catalog No. NB1001542

Supplier: Novus Biologicals NB1001542

Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Ymer Polyclonal specifically detects Ymer in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Ymer
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_848018.1
Gene Alias C3orf6deafness, autosomal dominant 44, coiled-coil domain containing 50, coiled-coil domain-containing protein 50, DFNA44, Protein Ymer, YMER
Gene Symbols CCDC50
Host Species Goat
Immunogen Peptide with sequence CFSKSESSHKGFHYK corresponding to internal region according to NP_848018.1, NP_777568.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152137
Test Specificity This antibody is expected to recognize both reported isoforms of human Ymer protein.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.