Learn More
Sigma Aldrich Fine Chemicals Biosciences X-tremeGENE 9 DNA Transfection Reagent, 0.4 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences 6365779001
X-tremeGENE 9 DNA Transfection Reagent is a non-liposomal multi-component reagent for experiments involving cellular analysis. Due to its extremely low cytotoxicity, minimal need for optimization, and the ability to provide high transfection efficiency in a wide range of commonly used cell lines even in the presence of serum, it is well suited for applications in all fields of cellular analysis.