Learn More
Electron Microscopy Sciences X-Checker™ Calibration Aid for SEM/EDS Standard
Supplier: Electron Microscopy Sciences 80058ST
The X-Checker was the first and remains the only complete calibration aid for SEM/EDS Systems. Each X-Checker comes with manganese to measure full width at half max detector resolution, aluminum and copper to perform spectral calibration, and carbon to monitor calibration at the low end of the spectra for thin window detectors. You also get two grid sizes for checking the accuracy of your image analysis software and an easy test for monitoring the amount of vacuum pump oil contamination on your detector window. Comes with: manganese, aluminum and copper, carbon.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.