Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Witch Hazel, USP, Spectrum™ Chemical
SDP

Catalog No. 18607747 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-747 500 mL Poly Bottle
18-607-748 4 L Poly Bottle
18-607-749 20 L Poly Drum
18-607-750 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-747 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. W1125500ML
Only null left
Add to Cart
Add to Cart

W1125

Spectrum™ Chemical Witch Hazel, USP is an astringent used externally on hemorrhoids, minor bleeding, and skin irritation. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS Max % 1
pH 3.0 to 5.0
Packaging Poly Bottle
Quantity 500 mL
Specific Gravity 0.979-0.983
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.