Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse WISP-1/CCN4 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 4 publications

$200.50 - $2109.88

Specifications

Antigen WISP-1/CCN4
Clone 214203
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1680SP
GREENER_CHOICE
View Documents
R&D Systems
MAB1680SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB1680100
GREENER_CHOICE
View Documents
R&D Systems
MAB1680100
100 μg
Each for $579.52
Only null left
Add to Cart
 
MAB1680500
GREENER_CHOICE
View Documents
R&D Systems
MAB1680500
500 μg
Each for $2,109.88
Only null left
Add to Cart
 
Description

Description

WISP-1/CCN4 Monoclonal specifically detects WISP-1/CCN4 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

WISP-1/CCN4
LYOPH
Western Blot
Unconjugated
Rat
Mouse
O54775
8840
Mouse myeloma cell line NS0-derived recombinant mouse WISP-1/CCN4 Leu21-Asn367 Accession # O54775
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse WISP-1/CCN4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 10-30% cross-reactivity with recombinant human (rh) WISP-1 but no cross-reactivity with rhCTGF or rmNOV.
214203
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CCN family member 4, CCN4, CCN4WISP1tc, WISP1, WISP-1, WISP1c, WISP1i, WNT1 induced secreted protein 1, Wnt-1 inducible signaling pathway protein 1, WNT1 inducible signaling pathway protein 1, wnt-1 signaling pathway protein 1, Wnt-1-induced secreted protein, WNT1-inducible-signaling pathway protein 1
WISP1
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.