Learn More
QIAGEN QIAcard FTA Wash Buffer
Catalog No. 09923348
Click to view available options
Quantity:
500 mL
Catalog No. 09-923-348
Supplier: QIAGEN WB120204
Only null left
Add to Cart
Add to Cart
For contamination-free collection, storage and transport of DNA samples.
QIAcard FTA Wash Buffer is used for purifying nucleic acids stored on FTA cards.
Specifications
For Use With (Equipment) QIAcard FTA cards, indicating FTA cards and EasiCollect device
For Use With (Application) Rapid purification of DNA from blood, cells, tissue, yeast, bacteria
Quantity 500 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.