Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QIAGEN QIAcard FTA Elute Micro

Catalog No. p-3170786
Click to view available options
Quantity:
100 Pk.
25 Pk.

Catalog No. 09-301-230

Supplier: QIAGEN WB120401

Only null left
Add to Cart
Add to Cart

For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.

  • FTA Elute technology for convenient water based elution of nucleic acids.
  • QIAcard FTA Elute Micro format with 4 spot area (spot volume 12 to 40 μL) to collect, stabilize, process, transport, and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.

Specifications

Content And Storage Room Temperature
Description Collect, process, transport and store colored biological samples, such as blood samples with Non-Indicating FTA Elute Micro Cards.
Sample Type Blood
For Use With (Application) Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection
No. of Samples 4
Product Type FTA Elute Micro Card
Quantity 25 Pk.
Salt Form Chaotropic
Sample Volume 40 μL
Test Time 15 to 30 min
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.