Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
What Seed Is It?

Click to view available options
:
Seed Identification kit
Description
Includes
- Nuts, pods, berries, acorns and other seeds to grow your own tree
- Seed key to identify unknown varieties
- Sprouting seed kit
Using 39 real tree seeds, students learn to key out, identify and classify by size, shape, color, type and means of transport.
- Grow sprouts and trees from seeds and study seed dispersal
- Each seed carded, labeled and bagged for protection
Specifications
Specifications
| Product Type | Seed Identification Kit |
| Grade | 3 to 12 |
| For Use With (Application) | Students key out, identify and classify real tree seeds by their size, shape, color, type and means of transport |
| Includes | 39 real tree seeds (each seed is carded, labeled and bagged), teacher's guide with hands-on activities, seed key for unknowns in kit, sprouting seed kit, seeds to grow your own tree, nuts, berries, pods, wings, acorns and more |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction