Learn More
Advansta Inc WESTERNBRIGHT ECL KIT, FOR 200
Supplier: Advansta Inc K12045D20
WesternBright ECL kit contains 200ml; sufficient for 2000cm2 membrane. Stronger signal than Amersham ECL or Pierce ECL. Detect low pg protein amounts. Use up to ten-times less antibody. WesternBright ECL is a horseradish peroxidase substrate optimized for chemiluminescent Western blots imaged using X-ray film. WesternBright ECL requires as little as ten times less antibody than other substrates, allowing you to save precious antibodies and samples. Additionally, the WesternBright ECL signal is long lasting, allowing multiple exposures to be carried out without substantial signal decay.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.