Learn More
LI COR BIOTECH LLC Black Western Blot Incubation Boxes (small - 5 boxes)
Supplier: LI COR BIOTECH LLC 92997105
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. The Small size is: 7.2 cm x 4.9 cm x 3.0 cm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.