Learn More
Chemglass Life Sciences Bottle, Washing, Safety, Polyethylene, 500mL, Acetone
Supplier: Chemglass Life Sciences CG113001
Bottle, Washing, Safety, Polyethylene, 500mL, Acetone. Easy-to-read, low-density polyethylene dispensing bottles are break-resistantIntended for short-term dispensing useInclude DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ informationEasy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information. Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label. Toluene bottle is solid red with white lettering and hazard information.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.