Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS45 Antibody, Novus Biologicals™
SDP

Catalog No. NB10041372 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10041372 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10041372 Supplier Novus Biologicals Supplier No. NB10041372
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

VPS45 Polyclonal specifically detects VPS45 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen VPS45
Applications Western Blot, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_009190.2
Gene Alias H1, H1VPS45, hlVps45, h-vps45, leucocyte vacuolar protein sorting 45, vacuolar protein sorting 45 homolog (S. cerevisiae), vacuolar protein sorting 45A, vacuolar protein sorting 45A (yeast homolog), vacuolar protein sorting 45A (yeast), vacuolar protein sorting-associated protein 45, VPS45A, VPS45AVPS54A, VPS45B, VSP45, VSP45A
Gene Symbols VPS45
Host Species Goat
Immunogen Peptide with sequence C-ESSQVTSRSASRR corresponding to C-Terminus according to NP_009190.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 11311
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Dog and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.