Learn More
Polyscience Viton Tubing 3/8 inch (9.5 mm) ID
SDP
Supplier:  Polyscience 060319
Encompass_Preferred
PolyScience™ Viton Tubing 3/8 (9.5 mm) ID-32 to 200°C3 ft (1 m) lengthsFor use with:• Circulating Baths• Refrigerated Circulating Baths• Heating Circulating Baths• Stainless Steel Open Bath Systems• Polycarbonate Open Bath Systems• Specialty Products• Viscosity Bath• Calibration Bath• 75L Refrigerated Circulating Bath• Chillers & Coolers• Benchtop Chiller
Catalog No. NC1898188
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.