Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Viral Production Medium

Catalog No. p-7232450
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
6 L
10 L
Catalog No. Quantity
A4817901 1 L
A4817902 6 L
A4817903 10 L
3 options

Catalog No. A4817901

Supplier: Gibco™ A4817901

Only null left
Add to Cart
Edge
Add to Cart

Viral Production Medium is a chemically defined, serum-free, and protein-free medium for growth and transfection of Viral Production Cells 2.0.

Gibco Viral Production Medium is a chemically defined, serum-free, and protein-free medium. It is animal origin-free and specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells).

  • For adeno-associated virus (AAV) production as part of the AAV-MAX Helper-Free AAV Production System
  • Supports growth of suspension Viral Production Cells 2.0 to densities of over 1.2 x 107 cells/mL
  • Enables production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Supports growth and transfection of Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Viral Production Medium requires supplementation with 4 mM GlutaMAX Supplement (Cat. No. 35050038) prior to use
TRUSTED_SUSTAINABILITY

Specifications

Cell Line Viral Production Cells 2.0
Cell Type HEK 293
Classification Serum-free, Animal Origin-free, Chemically-defined, Protein-free
Form Liquid
Product Type AAV Production Medium
Sterility Sterile-filtered
With Additives Glucose, Sodium Pyruvate
Without Additives No GlutaMAX
For Use With (Application) AAV Production
Manufacturing Quality ISO 13485, MDSAP, FDA-registered
Product Line AAV-MAX
Quantity 1 L
Shipping Condition Room Temperature
Format Bottle
pH 7
Regulatory Status RUO - Research Use Only
Content And Storage 1 L bottle; Reagents should be stored at 2°C to 8°C, protected from light
Culture Type Suspension Cell Culture
Expression System Mammalian
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.