Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Freudenberg™ Vileda™ QuickStar micro Microfiber Cloths
Shop All Vileda Professional Products
Click to view available options
Color:
Blue
Green
Red
Yellow
Description
- High chemical resistance against chlorine, bleach, disinfectants, peroxides, etc.
- Non-woven structure
- Easy to rinse clean, hairs and dirt do not cling to surface
- Very low-linting fabric due to patented endless fiber technology
- Launderable up to 300+ wash cycles
Specifications
Specifications
| Color | Red |
| Dimensions (L x W) | 16 x 16 in. |
| For Use With (Application) | ISO Class 7, LAR, Production, Labs |
| Packaging Quantity | 5 Cloths /Pk., 20 Pk./Cs. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction