Learn More
Vileda Professional™ UltraSpeed Pro Double Bucket System Ready To Go Kit
Catalog No. 19200718 Shop All Vileda Professional Products
Change view
Click to view available options
Capacity (English):
11 and 26 qt. Buckets
Catalog No. Capacity (English)
19-200-718 11 and 26 qt. Buckets
1 options
Catalog No. 19-200-718 Supplier Vileda Professional™ Supplier No. 148344
Developed to be simple to use, flexible enough to clean difficult areas and offer maximum cleaning performance while significantly reducing mopping times. Vileda Professional UltraSpeed Pro Double Bucket System Ready To Go Kit is a comprehensive floor cleaning system that sets new standards in usability, versatility, efficiency and ergonomics.
  • Ready to go kit includes two buckets with downpress wringer, chassis, push bar kit, extendable universal handle, 16”  UltraSpeed Pro frame and one UltraSpeed Trio mop
  • Double bucket system allows for separation of dirty liquid and clean liquid
  • Lightweight and robust buckets with ergonomic handles for carrying
  • Patented press: fastest and most efficient press on the market
  • Small and compact chassis
  • Changeable color coding clips within buckets
Specifications
Capacity (English) 11 and 26 qt. Buckets