Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VetMAX™-Gold AIV Detection Kit

The VetMAX™-Gold AIV Detection Kit (Avian Influenza Virus RNA Test Kit) is a highly sensitive, qualitative, one-step, real-time RT-PCR assay to detect Avian Influenza Virus (AIV) RNA isolated from poultry (chicken/turkey) oropharyngeal/tracheal swab samples.

Supplier:  LIFE TECHNOLOGIES 4485261

Encompass_Preferred

Catalog No. 44-852-61

  • $753.65 / Each of 1

    Save 5%

    Reg : $794.00

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

  • A highly sensitive, qualitative, one-step, real-time RT-PCR assay to detect Avian Influenza Virus (AIV) RNA isolated from poultry (chicken/turkey) oropharyngeal/tracheal swab samples
  • The VetMAX™-Gold AIV Detection Kit is a screening tool that detects sequences representing portions of the viral matrix (two distinct targets) and nucleoprotein (one target)
  • These are genes that are common to all AIV subtypes and found on viral RNA extracted from poultry samples
  • Assay is a single-well/tube, real-time RT-PCR assay in which RNA is reverse-transcribed into cDNA, and AIV and Xeno™ RNA targets are amplified and detected in real time using fluorescent TaqMan™ probes (hydrolysis probe chemistry)
  • Within the U.S., laboratories must receive authorization from their state veterinarian prior to ordering and using this kit for the diagnosis of myobacterium avium spp. paratuberculosis
  • Similar product use restrictions may exist in other countries, at the discretion of the relevant veterinary authority
  • Kit is not intended for differentiating AIV subtypes

Kit Includes:

  • Influenza Virus-Xeno™ RNA Control Mix: serves as a positive control for the real-time RT-PCR components and used to set the cycle threshold (CT) for evaluating test results
  • Xeno™ RNA Control: serves as an internal positive control for the RNA purification process and used to monitor for the presence of PCR inhibitors
  • Influenza Virus Primer Probe Mix: for optimized multiplex real-time RT-PCR amplification of Xeno™ RNA Control and AIV RNA targets
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2X Multiplex RT-PCR Buffer
Multiplex RT-PCR Enzyme Mix
Influenza Virus Primer Probe Mix
XenoTM RNA Control (10,000 copies/μL)
Influenza Virus-XenoTM RNA Control Mix (1000 copies/μL)
Nuclease-free Water

Store at -5 to -30°C.
Tube
qPCR
VetMAX™-Gold
100 reactions
Avian Influenza A
100 Reactions
Primer-probe
High
Avian Influenza
Real Time PCR Reagents
Avian
Animal Health
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Veterinary Use Only.