Learn More
Fine Science Tools Vannas Spring Scissors, Stainless Steel, 2.5mm Cutting Edge, 8 cm
Supplier: Fine Science Tools 1500208
Tip Shape: Angled to Side/ Tips: Sharp/ Cutting Edge: 2.5mm/ Tip Diameter: 0.075mm/ Alloy / Material: Stainless Steel/ Length: 8cm/ Serrated: No
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.