Learn More
Ace Glass, Inc. UV lamp, immersion, 200 watt, 121.92mm arc length, 250.95mmoverall length, metal-ended, no insulation, NEMA 15P power cord included
SDP
Supplier:  Ace Glass, Inc. 782532
Encompass
200watt med. pressure, quartz mercury-vapor lamp, for use in all ACE immersion wells. Arc length: 121.92mm. Overall length: 250.95mm. Supplied with 61cm PTFE covered lead wires, fitted with pin type connectors, and 6 cord to power supply.
Catalog No. 50-979-482