Learn More
Air Science Usa Air Science UV-BOX Benchtop Decontamination Chamber, Stainless Steel, 115V 60Hz
Supplier: Air Science Usa UVB15A
The UV-Box™ is a high-efficiency chamber designed to safely decontaminate contents while providing a safe work environment for the operator. High intensity UV lamps are positioned within the cabinet producing short wave ultraviolet light at 254 nm to destroy exposed surface DNA and bacteria, leaving evidence free of contamination prior to other forensic tests, analysis or procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.