Learn More
Greiner Bio-One Urine Set
Click to view available options
Bottom Shape:
Conical
Round
Description
- Additive: CCM (Count and Culture Mannitol) additive
- Tubes contain a stabilizer in powder form, which keeps the bacterial count in the sample stable at room temperature (20-25° C) for up to 48 hours
- The good solubility of the additive stabilizes the urine sample quickly and is therefore especially suitable in cases when the analysis takes place later
- This tube can be used as collection and/or transport container
Specifications
Specifications
| Closure Color | Yellow with Black Ring |
| Sterility | Non-sterile |
| Bottom Shape | Round |
| Description | Single-packed; Non-ridged tube; 13 x 75 |
| No. per Pack | 300 Pk. |
| Volume (Metric) | 4 mL |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov