Learn More
Eurofins Discoverx Uridine 5'-Triphosphate, 10 mg. (92-1109)
Supplier: Eurofins Discoverx 921109
Uridine 5'-Triphosphate (UTP) is a uracil nucleoside triphosphate that acts as a substrate for RNA synthesis during transcription. UTP also provides a source of energy and activates substrates in metabolic reactions, like ATP. Provided as 10 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter, PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.