Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™

Catalog No. BP169500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
This item is not returnable. View return policy

Catalog No. BP169-500

Only null left
Add to Cart
This item is not returnable. View return policy
Urea is used as a denaturing agent for DNA and proteins.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Absorbance 0.05 max. (5M solution) at 280nm
Melting Point 131°C
Color White
pH 7.5 to 9.5
Assay Percent Range ≥99 %
Packaging Poly Bottle
Linear Formula NH2CONH2
Quantity 500 g
Merck Index 15, 10052
Vapor Pressure 1.25mmHg at 25°C
Formula Weight 60.06
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Physical Form Solid
Chemical Name or Material Urea
Show More Show Less
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.