Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, ACS Reagent, Reagents

Catalog No. 01337634
Click to view available options
Quantity:
100 g
500 g

Catalog No. 01-337-634

Supplier: Reagents Holdings Llc C1345500100C2

Only null left
Add to Cart
Add to Cart

Urea, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Specifications

Boiling Range 135°C
Melting Point 133°C
Color White
Boiling Point 135°C
Melting Range 133°C
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 1.335
Grade ACS
Physical Form Solid
Chemical Name or Material Urea, ACS Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.