Learn More
Epredia™ UltraVision LP HRP Polymer & DAB Detection System
Shop All Epredia ProductsDescription
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: Diaminobenzidine (DAB)
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: Diaminobenzidine (DAB)
Includes:
TL-015-HD: 15mL Hydrogen Peroxide Block (TA-015-HP), 15mL Ultra V Block (TA-015-UB), 15mL Primary Antibody Enhancer (TL-015-PB), 15mL HRP Polymer (TL-015-PH), 15mL DAB Plus Substrate (TA-015-HSX), 1mL DAB Plus Chromogen (TA-001-HCX)
TL-060-HD: 60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Ultra V Block (TA-060-UB), 60mL Primary Antibody Enhancer (TL-060-PB), 60mL HRP Polymer (TL-060-PH), 60mL DAB Plus Substrate (TA-060-HSX), 2mL DAB Plus Chromogen (TA-002-HCX)
TL-125-HD: 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH), 125mL DAB Plus Substrate (TA-125-HSX), 4mL DAB Plus Chromogen (TA-004-HCX)
Note:
UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.
Specifications
Specifications
| Quantity | 15 mL |
| Count | 150 to 300 Slides |
| Includes | 15mL Hydrogen Peroxide Block (TA-015-HP), 15mL Ultra V Block (TA-015-UB), 15mL Primary Antibody Enhancer (TL-015-PB), 15mL HRP Polymer (TL-015-PH), 15mL DAB Plus Substrate (TA-015-HSX), 1mL DAB Plus Chromogen (TA-001-HCX) |
| Chromagen | Diaminobenzidine (DAB) |
| Specificity | anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
| Storage Requirements | Store at 2° to 8°C. Each component is stable for 18 months. |
| Enzyme | Peroxidase |
| Substrate | Diaminobenzidine (DAB) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.