Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UK 78282 hydrochloride
GREENER_CHOICE

Catalog No. 382950
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 38-295-0

Supplier: Tocris Bioscience™ 3829/50

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Blocker of KV1.3 and KV1.4 channels

Blocker of the KV1.3 and KV1.4 voltage-gated potassium channels in T lymphocytes (IC50 values are 0.28 and 0.17 μM respectively). Suppresses T lymphocyte mitogenesis; leads to membrane depolarization. Also blocks KV1.4 expressed in heart and brain.

Chemical Identifiers

CAS 136647-02-4
Molecular Formula C29H36ClNO2

Specifications

Quantity 50 mg
Formula Weight 466.05
Purity >99%
Molecular Weight (g/mol) 466.062
Chemical Name or Material UK 78282 hydrochloride
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.