Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ TRUFLOW™ Tissue and Biopsy Cassettes

Catalog No. p-3109749 Shop All Fisherbrand Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Product Type:
Biopsy Cassette
Tissue Cassette
This item is not returnable. View return policy

Catalog No. 15-200-403D

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Just bend, snap apart, and conveniently lock

  • Hold tissue and biopsy specimens during the embedding process as well as in a storage file
  • Molded from a special high-density polymer that is totally resistant to histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • Can be opened or closed as often as necessary and relocked securely without danger of specimen loss
  • Large labeling areas are located on three sides of the cassette
  • Anterior writing area at a 30° angle

Order Info

Packaged in three dispenser boxes of 500

Specifications

Pore Style Slots
Writing Face Angle 30°
Closure Type Integral Lid
Color Blue
Material High Density Polymer
Product Type Tissue Cassette
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.