Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAP100/MED24 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501560483
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
50-156-0483 100 μL
1 options

Catalog No. 50-156-0483

Supplier: Bethyl Laboratories, Inc A301472A

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Chicken

This gene encodes a component of the mediator complex , a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen TRAP100/MED24
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MED24
Gene Accession No. O75448, Q99K74
Gene Alias THRAP4, MED24, thyroid hormone receptor associated protein 4, ARC100, DRIP100, KIAA0130, MGC8748, TRAP100, activator-recruited cofactor 100 kDa component, mediator of RNA polymerase II transcription, subunit 24 homolog, thyroid hormone receptor-associate
Gene Symbols MED24
Host Species Rabbit
Immunogen Between 939 and 989
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23989, 9862
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.