Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-Dibenzylideneacetone, 98+%

Catalog No. AAA1209506
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AAA1209506

Supplier: Thermo Scientific Chemicals A1209506

Only null left
Add to Cart

CAS: 35225-79-7 | C17H14O | 234.30 g/mol

trans,trans-Dibenzylideneacetone was used as an additive in the copper-catalyzed-arylation of imidazoles. It is a reactant involved in Nazarov-like cyclization, Transfer hydrogenation, Lewis acid mediated condensation, Hetero-Diels-Alder reactions, Asymmetric 1,4-addition reactions and Michael addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-Dibenzylideneacetone was used as an additive in the copper-catalyzed-arylation of imidazoles. It is a reactant involved in Nazarov-like cyclization, Transfer hydrogenation, Lewis acid mediated condensation, Hetero-Diels-Alder reactions, Asymmetric 1,4-addition reactions and Michael addition reactions.

Solubility
Soluble in chloroform, acetone, alcohol (slightly), and hot methanol. Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35225-79-7
Molecular Formula C17H14O
Molecular Weight (g/mol) 234.30
MDL Number MFCD00004790
InChI Key WMKGGPCROCCUDY-PHEQNACWSA-N
Synonym dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one
PubChem CID 640180
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one
SMILES O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

Specifications

Melting Point 112°C to 114°C
Quantity 5 g
Beilstein 608434
Merck Index 14,3006
Solubility Information Soluble in chloroform,acetone,alcohol (slightly),and hot methanol. Insoluble in water.
Formula Weight 234.3
Percent Purity ≥98%
Chemical Name or Material trans,trans-Dibenzylideneacetone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.