Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Trace Minerals for an Ultra-Clean Environment

Catalog No. p-7113109 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g
500 g
1 kg

Catalog No. MP29602641

Supplier: MP Biomedicals™ 02960264.1

Only null left
Add to Cart
Add to Cart

Trace Minerals is used in ultra clean environments where trace elements usually present, are absent.

  • 7-trace elements which are usually present in a open environment
  • Research animal diet supplement
  • Powdered
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
For Use With (Application) Research animal diet supplements
Quantity 100 g
Storage Requirements Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.