Learn More
Stoelting™ Touch Test Sensory Probes
SDP
Catalog No. 10000250 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-250 Neuroscience
1 options
Catalog No. 10-000-250
Supplier: Stoelting™ 58011
Due to product restrictions, please to purchase or view availability for this product.
Semmes-Weinstein monofilaments provide an approximate logarithmic scale of actual force, and a linear scale of perceived intensity. They have a long history of effective use in clinical settings, and can be used to diagnose pathologies of hyper- or hypo-aesthesia. Subsets within the set of 20 probes distinguish pathologies on different parts of the body (foot, hand, lip, cheek,etc.).
Specifications
For Use With (Application) Neuroscience
Includes Kit of 20