Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ TissueLoc™ Cassette IV Slotted Cassettes in Tube Packs

Catalog No. p-7193541 Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow

Catalog No. B851720BE

Supplier: Epredia™ B851720BE

Only null left
Add to Cart
Add to Cart

Slotted Cassettes are ideal for use with routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in twelve color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving

Specifications

Pore Style Slots
Quantity 750/Cs.
Closure Included Yes
Closure Type Separate Lid, Back Hinge
PrintMate Compatible Yes
Color Beige
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type TissueLoc™ Slotted Cassette
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.