Learn More
NanoScreen LLC TIPS 30UL 384 50/CS B PE COMP
Supplier: NanoScreen LLC TIPP30384NBA
Pipette tips, Robotic. PerkinElmer P30, 384 tips. Non-Sterile, Aerosol Barrier. For PE/Packard 384 dispense heads found on MiniTrak, Janus and EP3. Tips are 100% inspected for quality. Guaranteed 1536 compatible. Case=50 racks=19200 tips
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.