Learn More
Labcon North America Eclipse™ 250uL Pipet Tips for Rainin LTS Pipettors, in 96 Racks, Sterile
Supplier: Labcon North America 10189600089
Tip, Pipet; Labcon; Eclipse™; Specialty; 250uL; Fits 250uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 960 tips per pack; 10 packs per case; 51.3mm. H; Made in USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.