Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ticarcillin disodium and clavulanate potassium (15:1)

Catalog No. AAJ6665909
Click to view available options
Quantity:
10 g
Packaging:
Plastic bottle

Catalog No. AAJ6665909

Supplier: Thermo Scientific Chemicals J6665909

Only null left
Add to Cart

CAS: 86482-18-0 | C23H25N3O11S2 | 583.583 g/mol

Combination provides greater efficacy against beta--lactam resistant strains

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86482-18-0
Molecular Formula C23H25N3O11S2
Molecular Weight (g/mol) 583.583
MDL Number MFCD26967928
InChI Key XWMVMWTVLSLJGY-FAJPTIRJSA-N
Synonym ticarcillin-clavulanic acid, clavulanic acid mixture with ticarcillin, clavulanic acid-ticarcillin mixt., ticarcillin-clavulanic acid mixt., ticarcillin disodium and clavulanate potassium, timentin in plastic container, 4-oxa-1-azabicyclo 3.2.0 heptane-2-carboxylic acid, 3-2-hydroxyethylidene-7-oxo-, 2r,3z,5r-, mixt. with 2s,5r,6r-6-2r-carboxy-3-thienylacetyl amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid, 4-oxa-1-azabicyclo 3.2.0 heptane-2-carboxylic acid, 3-2-hydroxyethylidene-7-oxo-, 2r-2-alpha,3z,5-alpha-, mixt. with 2s-2-alpha,5-alpha,6-beta s*-6-carboxy-3-thienylacetyl amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid, clavulanic acid-ticarcillin mixt
PubChem CID 6437075
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-carboxy-2-thiophen-3-ylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;(2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CSC=C3)C(=O)O)C(=O)O)C.C1C2N(C1=O)C(C(=CCO)O2)C(=O)O

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Freely soluble in water
Packaging Plastic bottle
Chemical Name or Material Ticarcillin disodium and clavulanate potassium (15:1)
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.