Learn More
Anaerobe Systems THIO 7 mL - Thioglycollate Broth 7 mL, 10 Tubes/Pkg
SDP
Supplier:  Anaerobe Systems AS801
Encompass
This item has a minimum qty of 3 per supplier requirements.
Thioglycollate Broth (THIO) is an enriched nonselective liquid medium used for the growth of microaerophilic and anaerobic bacteria, including fastidious organisms, from clinical specimens. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.THIO is an enriched nonselective liquid medium used for the growth of microaerophilic and fastidious anaerobic organisms. THIO contain casein, meat peptones, yeast extract, and dextrose as the nutritive base. Sodium thioglycollate and L-cystine are included as reducing agents. Hemin and vitamin K1 are included to promote the growth of fastidious anaerobes. A small quantity of agar is added to slow the diffusion of oxygen. A marble chip is included to each tube to act as a pH buffer and neutralize acids produced during growth of the organisms.
Catalog No. NC0174636
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.