Learn More
New England Biolabs, Inc. TelN Protelomerase – 250 units
Supplier: New England Biolabs, Inc. M0651S
TelN Protelomerase, from phage N15, cuts dsDNA at a TelN recognition sequence (56 bp) and leaves covalently closed ends at the site of cleavage.