Learn More
Ubpbio TCEP SOLUTION (1M), 100ML
Supplier: Ubpbio P1021100
Protein Purification Reagents – TCEP solution (1M) (UBPBio catalog# P1021-10, P1020-100) – TCEP (tris(2-carboxyethyl)phosphine) is a strong reducing agent. Compared to dithiothreitol (DTT) and β-mercaptoethanol, TCEP has the advantages of being odorless, a more powerful reducing agent, an irreversible reducing agent, more hydrophilic, and more resistant to oxidation in air. It is soluble in water and stable at neutral pH. It does not reduce metals used in immobilized metal affinity chromatography such as Ni-NTA resin. TCEP is particularly useful when labeling cysteine residues with maleimides.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.